Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzylmaleamic acid, 98+%

$34.35 - $342.04

Chemical Identifiers

Chemical Name or Material N-Benzylmaleamic acid
CAS 15329-69-8
Molecular Formula C11H11NO3
Formula Weight 205.21
MDL Number MFCD00020482
Beilstein 1212733
Synonym Maleic acid monobenzylamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2272606

  SDS
alfa aesar™
B2272606
5g Each for $34.35

AAB2272622

  SDS
alfa aesar™
B2272622
100g Each for $342.04
Description & Specifications

Specifications

Formula Weight 205.21
Chemical Name or Material N-Benzylmaleamic acid
Assay ≥98%
Molecular Formula C11H11NO3
Melting Point 136° to 138°C
CAS 15329-69-8
Beilstein 1212733
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00020482
Synonym Maleic acid monobenzylamide
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.