missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Benzyliminodiacetic Acid, TCI America™

$140.56 - $478.20

Chemical Identifiers

CAS 3987-53-9
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.228
MDL Number MFCD00004285
InChI Key SZQUPQVVCLFZLC-UHFFFAOYSA-N
PubChem CID 77606
IUPAC Name 2-[benzyl(carboxymethyl)amino]acetic acid
SMILES C1=CC=C(C=C1)CN(CC(=O)O)CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B13635G View Documents TCI America
B13635G
5g Each for $140.56
 
B136325G View Documents TCI America
B136325G
25g Each for $478.20
 
Specifications

Chemical Identifiers

3987-53-9
223.228
SZQUPQVVCLFZLC-UHFFFAOYSA-N
2-[benzyl(carboxymethyl)amino]acetic acid
C11H13NO4
MFCD00004285
77606
C1=CC=C(C=C1)CN(CC(=O)O)CC(=O)O

Specifications

White
5g
C11H13NO4
SZQUPQVVCLFZLC-UHFFFAOYSA-N
2-[benzyl(carboxymethyl)amino]acetic acid
77606
Crystal-Powder at 20°C
N-Benzyliminodiacetic Acid
201°C
3987-53-9
MFCD00004285
C1=CC=C(C=C1)CN(CC(=O)O)CC(=O)O
223.228
223.23
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : No