missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzylideneaniline, 99%

Manufacturer:  Alfa Aesar™ A1474614

 View more versions of this product

Catalog No. AAA1474614



Description

Description

Applications
N-Benzylideneaniline is an additional stain which is used in various immunoassays including flow cytometry, immunofluorescense, immunohistochemistry, and other applications.

Solubility
Soluble in water (partly), methanol, chloroform, alcohol, and acetic anhydride.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

N-Benzylideneaniline
52°C to 54°C
538-51-2
MFCD00003027
n-benzylideneaniline, benzalaniline, benzylideneaniline, e-n-benzylideneaniline, n-benzylidenaniline, n-benzalaniline, benzaldehyde anil, aniline, n-benzylidene, benzenamine, n-phenylmethylene, n-phenylmethylene benzenamine
C1=CC=C(C=C1)C=NC2=CC=CC=C2
181.238
181.24
>110°C (230°F)
25g
C13H11N
14,1138
UVEWQKMPXAHFST-UHFFFAOYSA-N
N,1-diphenylmethanimine
10858
99%

Chemical Identifiers

538-51-2
181.238
UVEWQKMPXAHFST-UHFFFAOYSA-N
10858
C1=CC=C(C=C1)C=NC2=CC=CC=C2
C13H11N
MFCD00003027
n-benzylideneaniline, benzalaniline, benzylideneaniline, e-n-benzylideneaniline, n-benzylidenaniline, n-benzalaniline, benzaldehyde anil, aniline, n-benzylidene, benzenamine, n-phenylmethylene, n-phenylmethylene benzenamine
N,1-diphenylmethanimine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents