Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzylidene-3,4-(methylenedioxy)benzylamine, 98+%

$67.11 - $915.86

Chemical Identifiers

Chemical Name or Material N-Benzylidene-3,4- (methylenedioxy)benzylamine
CAS 112776-37-1
Molecular Formula C15H13NO2
Formula Weight 239.3
MDL Number MFCD00115461
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4453403

  SDS
alfa aesar™
4453403
1g Each for $67.11

AA4453414

  SDS
alfa aesar™
4453414
25g Each for $915.86
Description & Specifications

Specifications

Formula Weight 239.3
Chemical Name or Material N-Benzylidene-3,4- (methylenedioxy)benzylamine
Physical Form Powder
Assay ≥98%
Molecular Formula C15H13NO2
Melting Point 69° to 70°C
MDL Number MFCD00115461
CAS 112776-37-1
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.