missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzylidene-3,4-(methylenedioxy)benzylamine, 98+%

Manufacturer:  Alfa Aesar™ 4453414

 View more versions of this product

Catalog No. AA4453414



Specifications

Specifications

N-Benzylidene-3,4- (methylenedioxy)benzylamine
25g
C15H13NO2
239.3
≥98%
69°C to 70°C
112776-37-1
MFCD00115461
Powder

Chemical Identifiers

112776-37-1
C15H13NO2
MFCD00115461
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents