missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Benzylhydroxylamine Hydrochloride, TCI America™

$155.84 - $556.13

Chemical Identifiers

CAS 29601-98-7
Molecular Formula C7H10ClNO
Molecular Weight (g/mol) 159.613
MDL Number MFCD00043377
InChI Key YSNXOQGDHGUKCZ-UHFFFAOYSA-N
Synonym n-benzylhydroxylamine hydrochloride, n-benzylhydroxylamine hcl, n-benzyl-n-hydroxylamine hydrochloride, benzenemethanamine, n-hydroxy-, hydrochloride, n-benzylhydroxyamine hydrochloride, bnnhoh*hydrochloride;, acmc-1cr2p, ksc493k8t, benzylhydroxyamine hydrochloride, n-benzylhydroxylamne hydrochlorde
PubChem CID 11332622
IUPAC Name N-benzylhydroxylamine;hydrochloride
SMILES C1=CC=C(C=C1)CNO.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B40735G View Documents TCI America
B40735G
5g Each for $155.84
 
B407325G View Documents TCI America
B407325G
25g Each for $556.13
 
Specifications

Chemical Identifiers

29601-98-7
159.613
YSNXOQGDHGUKCZ-UHFFFAOYSA-N
11332622
C1=CC=C(C=C1)CNO.Cl
C7H10ClNO
MFCD00043377
n-benzylhydroxylamine hydrochloride, n-benzylhydroxylamine hcl, n-benzyl-n-hydroxylamine hydrochloride, benzenemethanamine, n-hydroxy-, hydrochloride, n-benzylhydroxyamine hydrochloride, bnnhoh*hydrochloride;, acmc-1cr2p, ksc493k8t, benzylhydroxyamine hydrochloride, n-benzylhydroxylamne hydrochlorde
N-benzylhydroxylamine;hydrochloride

Specifications

White
5g
C7H10ClNO
n-benzylhydroxylamine hydrochloride, n-benzylhydroxylamine hcl, n-benzyl-n-hydroxylamine hydrochloride, benzenemethanamine, n-hydroxy-, hydrochloride, n-benzylhydroxyamine hydrochloride, bnnhoh*hydrochloride;, acmc-1cr2p, ksc493k8t, benzylhydroxyamine hydrochloride, n-benzylhydroxylamne hydrochlorde
C1=CC=C(C=C1)CNO.Cl
159.613
159.61
≥98.0% (HPLC,T)
106°C
29601-98-7
MFCD00043377
YSNXOQGDHGUKCZ-UHFFFAOYSA-N
N-benzylhydroxylamine;hydrochloride
11332622
Crystal-Powder at 20°C
N-Benzylhydroxylamine Hydrochloride
Safety and Handling

Safety and Handling

TSCA : No