missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Benzylglycine Hydrochloride, TCI America™

$24.44 - $68.75

Chemical Identifiers

CAS 7689-50-1
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD00156934
InChI Key BUZJPENZWLUHJD-UHFFFAOYSA-N
Synonym n-benzylglycine hydrochloride, benzylaminoacetic acid hydrochloride, 2-benzylamino acetic acid hydrochloride, bzl-gly-oh.hcl, n-benzylglycine hcl, bz-gly-oh.hcl, n-benzyl glycine hydrochloride, bzl-gly-oh inverted exclamation mark currencyhcl, benzylamino acetic acid hydrochloride, glycine, n-phenylmethyl-, hydrochloride
PubChem CID 2756624
IUPAC Name 2-(benzylamino)acetic acid;hydrochloride
SMILES C1=CC=C(C=C1)CNCC(=O)O.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Qty  
B30755G missing translation for 'viewDocuments' TCI America
B30755G
5g Each for $24.44
 
B307525G missing translation for 'viewDocuments' TCI America
B307525G
25g Each for $68.75
 
Specifications

Chemical Identifiers

7689-50-1
201.65
BUZJPENZWLUHJD-UHFFFAOYSA-N
2756624
C1=CC=C(C=C1)CNCC(=O)O.Cl
C9H12ClNO2
MFCD00156934
n-benzylglycine hydrochloride, benzylaminoacetic acid hydrochloride, 2-benzylamino acetic acid hydrochloride, bzl-gly-oh.hcl, n-benzylglycine hcl, bz-gly-oh.hcl, n-benzyl glycine hydrochloride, bzl-gly-oh inverted exclamation mark currencyhcl, benzylamino acetic acid hydrochloride, glycine, n-phenylmethyl-, hydrochloride
2-(benzylamino)acetic acid;hydrochloride

Specifications

White-Yellow
5g
C9H12ClNO2
n-benzylglycine hydrochloride, benzylaminoacetic acid hydrochloride, 2-benzylamino acetic acid hydrochloride, bzl-gly-oh.hcl, n-benzylglycine hcl, bz-gly-oh.hcl, n-benzyl glycine hydrochloride, bzl-gly-oh inverted exclamation mark currencyhcl, benzylamino acetic acid hydrochloride, glycine, n-phenylmethyl-, hydrochloride
C1=CC=C(C=C1)CNCC(=O)O.Cl
201.65
201.65
≥98.0% (HPLC)
228°C
7689-50-1
MFCD00156934
BUZJPENZWLUHJD-UHFFFAOYSA-N
2-(benzylamino)acetic acid;hydrochloride
2756624
Crystal-Powder at 20°C
N-Benzylglycine Hydrochloride
Safety and Handling

Safety and Handling

TSCA : No