Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzylglycine hydrochloride, 98+%

$52.03 - $547.99

Chemical Identifiers

Chemical Name or Material N-Benzylglycine hydrochloride
CAS 7689-50-1
Molecular Formula C6H5CH2NHCH2CO2H·HCl
Formula Weight 201.65
MDL Number MFCD00156934
Beilstein 3915652
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1520006

  SDS
alfa aesar™
L1520006
5g Each for $52.03

AAL1520014

  SDS
alfa aesar™
L1520014
25g Each for $150.85

AAL1520022

  SDS
alfa aesar™
L1520022
100g Each for $547.99
Description & Specifications

Specifications

Formula Weight 201.65
Molecular Formula C6H5CH2NHCH2CO2H·HCl
MDL Number MFCD00156934
CAS 7689-50-1
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material N-Benzylglycine hydrochloride
Assay ≥98%
Beilstein 3915652
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈220°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.