missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Benzylglycine Ethyl Ester 96.0+%, TCI America™
Supplier: TCI America B227325G
Specifications
N-Benzylglycine Ethyl Ester | |
Yellow | |
C11H15NO2 | |
25 g | |
ULOLIZHBYWAICY-UHFFFAOYSA-N | |
ethyl 2-(benzylamino)acetate | |
80908 | |
≥96.0% (GC) |
6436-90-4 | |
142°C | |
MFCD00009174 | |
n-benzylglycine ethyl ester, ethyl 2-benzylamino acetate, ethyl n-benzylglycinate, n-bzl-gly-oet, bzl-gly-oet, glycine, n-phenylmethyl-, ethyl ester, n-benzyl glycine ethyl ester, ethyl n-phenylmethyl glycinate, ethyl benzylamino acetate, n-benzyl-glycine ethyl ester | |
CCOC(=O)CNCC1=CC=CC=C1 | |
193.25 | |
193.25 | |
Liquid |
Chemical Identifiers
6436-90-4 | |
193.25 | |
ULOLIZHBYWAICY-UHFFFAOYSA-N | |
80908 | |
CCOC(=O)CNCC1=CC=CC=C1 |
C11H15NO2 | |
MFCD00009174 | |
n-benzylglycine ethyl ester, ethyl 2-benzylamino acetate, ethyl n-benzylglycinate, n-bzl-gly-oet, bzl-gly-oet, glycine, n-phenylmethyl-, ethyl ester, n-benzyl glycine ethyl ester, ethyl n-phenylmethyl glycinate, ethyl benzylamino acetate, n-benzyl-glycine ethyl ester | |
ethyl 2-(benzylamino)acetate |
Safety and Handling
EINECSNumber : (3)-1723
TSCA : No
Recommended Storage : Refrigerator