missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Benzylethylenediamine, TCI America™

$71.81 - $203.20

Chemical Identifiers

CAS 4152-09-4
Molecular Formula C9H14N2
Molecular Weight (g/mol) 150.23
MDL Number MFCD00041896
InChI Key ACYBVNYNIZTUIL-UHFFFAOYSA-N
Synonym n-benzylethylenediamine, n1-benzylethane-1,2-diamine, 2-aminoethyl benzyl amine, ethylenediamine, n-benzyl, 1,2-ethanediamine, n-phenylmethyl, n-2-aminoethyl-n-benzylamine, 2-benzylaminoethylamine, n-benzylethane-1,2-diamine, 1,2-ethanediamine, n1-phenylmethyl, 2-aminoethyl benzylamine
PubChem CID 77801
IUPAC Name N'-benzylethane-1,2-diamine
SMILES C1=CC=C(C=C1)CNCCN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B11065ML View Documents TCI America
B11065ML
5mL Each for $71.81
 
B110625ML View Documents TCI America
B110625ML
25mL Each for $203.20
 
Specifications

Chemical Identifiers

4152-09-4
150.23
ACYBVNYNIZTUIL-UHFFFAOYSA-N
77801
C1=CC=C(C=C1)CNCCN
C9H14N2
MFCD00041896
n-benzylethylenediamine, n1-benzylethane-1,2-diamine, 2-aminoethyl benzyl amine, ethylenediamine, n-benzyl, 1,2-ethanediamine, n-phenylmethyl, n-2-aminoethyl-n-benzylamine, 2-benzylaminoethylamine, n-benzylethane-1,2-diamine, 1,2-ethanediamine, n1-phenylmethyl, 2-aminoethyl benzylamine
N'-benzylethane-1,2-diamine

Specifications

165°C
5mL
C9H14N2
2735
ACYBVNYNIZTUIL-UHFFFAOYSA-N
N'-benzylethane-1,2-diamine
77801
Clear Liquid at 20°C
N-Benzylethylenediamine
Yellow
4152-09-4
MFCD00041896
n-benzylethylenediamine, n1-benzylethane-1,2-diamine, 2-aminoethyl benzyl amine, ethylenediamine, n-benzyl, 1,2-ethanediamine, n-phenylmethyl, n-2-aminoethyl-n-benzylamine, 2-benzylaminoethylamine, n-benzylethane-1,2-diamine, 1,2-ethanediamine, n1-phenylmethyl, 2-aminoethyl benzylamine
C1=CC=C(C=C1)CNCCN
150.23
150.23
≥97.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0312&(9)-1312

TSCA : Yes