Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzylethylenediamine, 98%

Chemical Identifiers

Chemical Name or Material N-Benzylethylenediamine
CAS 4152-09-4
Molecular Formula C6H5CH2NHCH2CH2NH2
Formula Weight 150.23
MDL Number MFCD00041896
UN Number UN2735
Beilstein 907505
Synonym N-(2-Aminoethyl)benzylamine; 2-(Benzylamino)ethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1326288

  SDS
alfa aesar™
A1326288
2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1326209

  SDS
alfa aesar™
A1326209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1326218

  SDS
alfa aesar™
A1326218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material N-Benzylethylenediamine
CAS 4152-09-4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA TSCA
Assay 98%
Molecular Formula C6H5CH2NHCH2CH2NH2
Formula Weight 150.23
MDL Number MFCD00041896
UN Number UN2735
Boiling Point 162°C (20mmHg)
Beilstein 907505
Quantity 2.5g
Flash Point 95°C (203°F)
Synonym N-(2-Aminoethyl)benzylamine; 2-(Benzylamino)ethylamine
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.