Learn More
N-Benzyldimethylamine, 98+%, Thermo Scientific Chemicals
$70.01 - $749.95
Chemical Identifiers
CAS | 103-83-3 |
---|---|
Molecular Formula | C9H13N |
Molecular Weight (g/mol) | 135.21 |
MDL Number | MFCD00008329 |
InChI Key | XXBDWLFCJWSEKW-UHFFFAOYSA-N |
Synonym | n,n-dimethylbenzylamine, benzyldimethylamine, n-benzyldimethylamine, dimethylbenzylamine, bdma, benzenemethanamine, n,n-dimethyl, benzyl-n,n-dimethylamine, n-phenylmethyl dimethylamine, n,n-dimethylbenzenemethanamine, araldite accelerator 062 |
PubChem CID | 7681 |
IUPAC Name | N,N-dimethyl-1-phenylmethanamine |
SMILES | CN(C)CC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA12191AK
|
Thermo Scientific Chemicals
A12191AK |
250 mL |
N/A
|
|
|||||
AAA12191AU
|
Thermo Scientific Chemicals
A12191AU |
1000 mL |
N/A
|
|
|||||
AAA121910J
|
Thermo Scientific Chemicals
A121910J |
5000 mL |
N/A
|
|
|||||
Description
N,N-Dimethylbenzylamine is used in the preparation of bis[(N,N-dimethylamino)benzyl] selenide. It acts as a catalyst in the curing reaction of formulations of diglycidyl ether of bisphenol A and tetrahydrophthalic anhydride. It undergoes directed ortho metalation with butyl lithium. It reacts with methyl iodide to get ammonium salt, which is used as phase transfer catalysts. Further, it is used as a catalyst for the formation of polyurethane foams and epoxy resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N-Dimethylbenzylamine is used in the preparation of bis[(N,N-dimethylamino)benzyl] selenide. It acts as a catalyst in the curing reaction of formulations of diglycidyl ether of bisphenol A and tetrahydrophthalic anhydride. It undergoes directed ortho metalation with butyl lithium. It reacts with methyl iodide to get ammonium salt, which is used as phase transfer catalysts. Further, it is used as a catalyst for the formation of polyurethane foams and epoxy resins.
Solubility
Slightly miscible with water.
Notes
Air sensitive. Incompatible with strong acids, strong bases, strong oxidizing agents, strong reducing agents and carbon dioxide.
Chemical Identifiers
103-83-3 | |
135.21 | |
XXBDWLFCJWSEKW-UHFFFAOYSA-N | |
7681 | |
CN(C)CC1=CC=CC=C1 |
C9H13N | |
MFCD00008329 | |
n,n-dimethylbenzylamine, benzyldimethylamine, n-benzyldimethylamine, dimethylbenzylamine, bdma, benzenemethanamine, n,n-dimethyl, benzyl-n,n-dimethylamine, n-phenylmethyl dimethylamine, n,n-dimethylbenzenemethanamine, araldite accelerator 062 | |
N,N-dimethyl-1-phenylmethanamine |
Specifications
103-83-3 | |
0.9 | |
54°C (129°F) | |
1.501 | |
MFCD00008329 | |
UN2619 | |
Air sensitive | |
Slightly miscible with water. | |
CN(C)CC1=CC=CC=C1 | |
135.21 | |
135.21 | |
N-Benzyldimethylamine |
-75°C | |
183°C to 184°C | |
C9H13N | |
C6H5CH2N(CH3)2 | |
250 mL | |
1099620 | |
n,n-dimethylbenzylamine, benzyldimethylamine, n-benzyldimethylamine, dimethylbenzylamine, bdma, benzenemethanamine, n,n-dimethyl, benzyl-n,n-dimethylamine, n-phenylmethyl dimethylamine, n,n-dimethylbenzenemethanamine, araldite accelerator 062 | |
XXBDWLFCJWSEKW-UHFFFAOYSA-N | |
N,N-dimethyl-1-phenylmethanamine | |
7681 | |
≥98% |
Safety and Handling
GHS H Statement
H314-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H302+H312-H314-H331-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: BENZYLDIMETHYLAMINE
EINECSNumber : 203-149-1
RTECSNumber : DP4500000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only