missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyldimethylamine, 98+%

Manufacturer:  Alfa Aesar™ A12191AK

 View more versions of this product

Catalog No. AAA12191AK


 

Description

Description

Applications
N,N-Dimethylbenzylamine is used in the preparation of bis[(N,N-dimethylamino)benzyl] selenide. It acts as a catalyst in the curing reaction of formulations of diglycidyl ether of bisphenol A and tetrahydrophthalic anhydride. It undergoes directed ortho metalation with butyl lithium. It reacts with methyl iodide to get ammonium salt, which is used as phase transfer catalysts. Further, it is used as a catalyst for the formation of polyurethane foams and epoxy resins.

Solubility
Slightly miscible with water.

Notes
Air sensitive. Incompatible with strong acids, strong bases, strong oxidizing agents, strong reducing agents and carbon dioxide.
Specifications

Specifications

N-Benzyldimethylamine
≥98%
C6H5CH2N(CH3)2
-75°C
1099620
250mL
N,N-Dimethylbenzylamine
CN(C)CC1=CC=CC=C1
135.21
135.21
103-83-3
C9H13N
MFCD00008329
UN2619
183°C to 184°C
54°C (129°F)
XXBDWLFCJWSEKW-UHFFFAOYSA-N
N,N-dimethyl-1-phenylmethanamine
7681

Chemical Identifiers

103-83-3
135.21
XXBDWLFCJWSEKW-UHFFFAOYSA-N
7681
C9H13N
MFCD00008329
N,N-Dimethylbenzylamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: BENZYLDIMETHYLAMINE

RTECSNumber : DP4500000

TSCA : TSCA

SDS Documents