Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzylbenzamide, 99+%

$97.12 - $281.69

Chemical Identifiers

Chemical Name or Material N-Benzylbenzamide
CAS 1485-70-7
Molecular Formula C6H5CONHCH2C6H5
Formula Weight 211.26
MDL Number MFCD00003070
Beilstein 2211315
Synonym N-Benzoylbenzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1220414

  SDS
alfa aesar™
A1220414
25g Each for $97.12

AAA1220422

  SDS
alfa aesar™
A1220422
100g Each for $281.69
Description & Specifications

Specifications

Formula Weight 211.26
Chemical Name or Material N-Benzylbenzamide
Assay ≥99%
Molecular Formula C6H5CONHCH2C6H5
MDL Number MFCD00003070
Beilstein 2211315
Quantity 25g
Synonym N-Benzoylbenzylamine
Melting Point 104° to 108°C
CAS 1485-70-7

Applications
N-Benzylbenzamide is a convenient precursor to α-substituted benzyl amines and an indicator for the titration of butyl lithium and other lithium bases.

Solubility
Soluble in Acetone (25 mg/ml).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.