Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzylaminoacetonitrile hydrochloride, 98%

$74.34 - $74.34

Chemical Identifiers

Chemical Name or Material N-Benzylaminoacetonitrile hydrochloride
CAS 63086-36-2
Molecular Formula C6H5CH2NHCH2CN·HCl
Formula Weight 182.65
MDL Number MFCD02094033
UN Number UN3439
Beilstein 3699545
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1856606

 
alfa aesar™
L1856606*
5g Each for $74.34

AAL1856614

 
alfa aesar™
L1856614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 182.65
Chemical Name or Material N-Benzylaminoacetonitrile hydrochloride
Assay 98%
Molecular Formula C6H5CH2NHCH2CN·HCl
Melting Point 153° to 160°C
MDL Number MFCD02094033
CAS 63086-36-2
UN Number UN3439
Beilstein 3699545
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.