Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzylaminoacetaldehyde diethyl acetal, 96%

$527.36 - $527.36

Chemical Identifiers

Chemical Name or Material N-Benzylaminoacetaldehyde diethyl acetal
CAS 61190-10-1
Molecular Formula C6H5CH2NHCH2CH(OCH2CH3)2
Formula Weight 223.32
MDL Number MFCD00051599
Beilstein 2694610
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1384114

  SDS
alfa aesar™
A1384114
25g Each for $527.36
Description & Specifications

Specifications

Formula Weight 223.32
Beilstein 2694610
MDL Number MFCD00051599
Quantity 25g
Assay 96%
Boiling Point 115° to 117°C (1mmHg)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 61190-10-1
Chemical Name or Material N-Benzylaminoacetaldehyde diethyl acetal
Molecular Formula C6H5CH2NHCH2CH(OCH2CH3)2
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.