missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyl-N-tert-butyl-4-(chloromethyl)benzamide, 97%

Chemical Identifiers

CAS 1448636-17-6
Molecular Formula C19H22ClNO
Molecular Weight (g/mol) 315.841
MDL Number MFCD22683147
InChI Key AGBSCAUSQCMREP-UHFFFAOYSA-N
Synonym n-benzyl-n-tert-butyl-4-chloromethyl benzamide
PubChem CID 98000581
IUPAC Name N-benzyl-N-tert-butyl-4-(chloromethyl)benzamide
SMILES CC(C)(C)N(CC1=CC=CC=C1)C(=O)C2=CC=C(C=C2)CCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60751MD View Documents Alfa Aesar™
H60751MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6075103 View Documents Alfa Aesar™
H6075103
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1448636-17-6
315.841
AGBSCAUSQCMREP-UHFFFAOYSA-N
98000581
CC(C)(C)N(CC1=CC=CC=C1)C(=O)C2=CC=C(C=C2)CCl
C19H22ClNO
MFCD22683147
n-benzyl-n-tert-butyl-4-chloromethyl benzamide
N-benzyl-N-tert-butyl-4-(chloromethyl)benzamide

Specifications

250mg
C19H22ClNO
n-benzyl-n-tert-butyl-4-chloromethyl benzamide
CC(C)(C)N(CC1=CC=CC=C1)C(=O)C2=CC=C(C=C2)CCl
315.841
315.84
N-Benzyl-N-tert-butyl-4-(chloromethyl)benzamide
1448636-17-6
MFCD22683147
AGBSCAUSQCMREP-UHFFFAOYSA-N
N-benzyl-N-tert-butyl-4-(chloromethyl)benzamide
98000581
97%