missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyl-N,N'-dimethylethylenediamine 98.0+%, TCI America™
$275.87 - $935.76
Chemical Identifiers
| CAS | 102-11-4 |
|---|---|
| Molecular Formula | C11H18N2 |
| Molecular Weight (g/mol) | 178.28 |
| MDL Number | MFCD00014856 |
| InChI Key | PVFIKWLTVKSXED-UHFFFAOYSA-N |
| PubChem CID | 66016 |
| IUPAC Name | benzyl(methyl)[2-(methylamino)ethyl]amine |
| SMILES | CNCCN(C)CC1=CC=CC=C1 |
Chemical Identifiers
| 102-11-4 | |
| 178.28 | |
| PVFIKWLTVKSXED-UHFFFAOYSA-N | |
| benzyl(methyl)[2-(methylamino)ethyl]amine |
| C11H18N2 | |
| MFCD00014856 | |
| 66016 | |
| CNCCN(C)CC1=CC=CC=C1 |
Specifications
| 102-11-4 | |
| 74°C | |
| MFCD00014856 | |
| PVFIKWLTVKSXED-UHFFFAOYSA-N | |
| benzyl(methyl)[2-(methylamino)ethyl]amine | |
| 66016 | |
| ≥98.0% (GC,T) | |
| N-Benzyl-N,N′-dimethylethylenediamine |
| Yellow | |
| C11H18N2 | |
| 5 g | |
| CNCCN(C)CC1=CC=CC=C1 | |
| 178.28 | |
| 178.28 | |
| Liquid |
Safety and Handling
TSCA : Yes