Please login to your online account to display your discounted pricing

Alfa Aesar™ N'-Benzyl-N,N-dimethylethylenediamine, 97%

Chemical Identifiers

Chemical Name or Material N'-Benzyl-N,N-dimethylethylenediamine
CAS 103-55-9
Molecular Formula C11H18N2
Linear Formula C6H5CH2NHCH2CH2N(CH3)2
Formula Weight 178.28
MDL Number MFCD00008334
UN Number UN2735
Beilstein 2641339
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0312606

 
alfa aesar™
L0312606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0312614

 
alfa aesar™
L0312614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 178.28
Chemical Name or Material N'-Benzyl-N,N-dimethylethylenediamine
Molecular Formula C11H18N2
CAS 103-55-9
TSCA TSCA
Assay 97%
Linear Formula C6H5CH2NHCH2CH2N(CH3)2
MDL Number MFCD00008334
UN Number UN2735
Beilstein 2641339
Quantity 5g
Odor Amine-like
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Boiling Point 255°C
Flash Point >110°C (230°F)
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.