Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyl-N-ethylaniline, 98%

Chemical Identifiers

Chemical Name or Material N-Benzyl-N-ethylaniline
CAS 92-59-1
Molecular Formula C15H17N
Formula Weight 211.31
MDL Number MFCD00009037
UN Number UN2274
Beilstein 2099852
Merck Index 14,3769
Synonym N-Ethyl-N-phenylbenzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1988209

  SDS
alfa aesar™
A1988209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1988218

  SDS
alfa aesar™
A1988218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1988230

  SDS
alfa aesar™
A1988230
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 211.31
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: N-ETHYL-N-BENZYLANILINE
Quantity 10g
Flash Point >157°C (315°F)
Synonym N-Ethyl-N-phenylbenzylamine
CAS 92-59-1
MDL Number MFCD00009037
Boiling Point 163° to 164°C (6mmHg)
Beilstein 2099852
Health Hazard 2 GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
UN Number UN2274
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material N-Benzyl-N-ethylaniline
Merck Index 14,3769
Assay 98%
TSCA TSCA
Molecular Formula C15H17N
Safety and Handling

Warning

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.