Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyl-n-butylamine, 98%

$45.81 - $45.81

Chemical Identifiers

Chemical Name or Material N-Benzyl-n-butylamine
CAS 2403-22-7
Molecular Formula C6H5CH2NH(CH2)3CH3
Formula Weight 163.27
MDL Number MFCD00009427
UN Number UN2735
Beilstein 2081422
Synonym N-n-Butylbenzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1785106

  SDS
alfa aesar™
A1785106
5g Each for $45.81

AAA1785114

  SDS
alfa aesar™
A1785114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 163.27
TSCA TSCA
UN Number UN2735
Beilstein 2081422
Quantity 5g
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula C6H5CH2NH(CH2)3CH3
Flash Point 94°C (201°F)
MDL Number MFCD00009427
Assay 98%
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 87° to 89°C (3mmHg)
Health Hazard 1 Danger
Chemical Name or Material N-Benzyl-n-butylamine
Synonym N-n-Butylbenzylamine
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
CAS 2403-22-7
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.