missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyl-N-(3-pyridylmethylene)amine

$78.04 - $273.99

Chemical Identifiers

CAS 71718-88-2
Molecular Formula C13H12N2
Formula Weight 196.25
MDL Number MFCD08064239
Chemical Name or Material N-Benzyl-N-(3-pyridylmethylene)amine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5074503 SDS Alfa Aesar™
H5074503
1g Each for $273.99
AAH50745MD SDS Alfa Aesar™
H50745MD
250mg Each for $78.04
Specifications Description & Specifications

Chemical Identifiers

71718-88-2
196.25
N-Benzyl-N-(3-pyridylmethylene)amine
C13H12N2
MFCD08064239

Specifications

71718-88-2
196.25
1g
C13H12N2
MFCD08064239
N-Benzyl-N-(3-pyridylmethylene)amine

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning