Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyl-beta-alanine ethyl ester, 95%

$24.95 - $136.38

Chemical Identifiers

Chemical Name or Material N-Benzyl-beta-alanine ethyl ester
CAS 23583-21-3
Molecular Formula C12H17NO2
Linear Formula C6H5CH2NHCH2CH2CO2CH2CH3
Formula Weight 207.27
MDL Number MFCD00085541
Synonym Bzl-beta-Ala-OEt; Ethyl 3-(benzylamino)propionate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1765203

 
alfa aesar™
L1765203
1g Each for $24.95

AAL1765206

 
alfa aesar™
L1765206
5g Each for $42.19

AAL1765214

 
alfa aesar™
L1765214
25g Each for $136.38
Description & Specifications

Specifications

Formula Weight 207.27
Chemical Name or Material N-Benzyl-beta-alanine ethyl ester
Assay 98%
Molecular Formula C12H17NO2
Linear Formula C6H5CH2NHCH2CH2CO2CH2CH3
MDL Number MFCD00085541
CAS 23583-21-3
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Synonym Bzl-beta-Ala-OEt; Ethyl 3-(benzylamino)propionate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Boiling Point 150°C (0.2mmHg)
Quantity 1g
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

2° to 8°C