Please login to your online account to display your discounted pricing

N-Benzyl-4-piperidone, 99%, ACROS Organics™


Quantity 250mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  106132500

 View more versions of this product

Catalog No. AC106132500

  • / Each
 

Description & Specifications

Chemical Identifiers

Color Clear colorless to straw
Physical Form Oily Liquid
Formula Weight 189.26
Quantity 250mL
Assay 98 to 102%
EINECS Number 222-782-4
CAS Max % 100.0
Chemical Name or Material N-Benzyl-4-piperidone
Assay Percent Range 99%
Name Note 99%
MDL Number MFCD00006192
CAS 3612-20-2
Molecular Formula C12H15NO
CAS Min % 98.5
Refractive Index 1.5390 to 1.5410 (20°C, 589nm)
Infrared Spectrum Authentic
Storage Information This product gains color during storage
Water 0.5% max.
Packaging Glass bottle
Synonym 1-(Phenylmethyl)-4-piperidinone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material N-Benzyl-4-piperidone
Name Note 99%
CAS 3612-20-2
CAS Min % 98.5
CAS Max % 100.0
Assay 98 to 102%
Assay Percent Range 99%
Molecular Formula C12H15NO
Formula Weight 189.26
MDL Number MFCD00006192
Synonym 1-(Phenylmethyl)-4-piperidinone
Accessories

Warning

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.

SDS & Safety Documents