Learn More
N-Benzyl-4-piperidone, 99%, ACROS Organics™
$139.21 - $281.65
Chemical Identifiers
CAS | 3612-20-2 |
---|---|
Molecular Formula | C12H15NO |
Molecular Weight (g/mol) | 189.26 |
MDL Number | MFCD00006192 |
InChI Key | SJZKULRDWHPHGG-UHFFFAOYSA-N |
Synonym | 1-benzyl-4-oxopiperidine, 1-benzyl-4-piperidinone, 1-benzyl-4-piperidone, 1-benzyl-piperidin-4-one, 1-benzylpiperidone, 4-piperidinone, 1-phenylmethyl, n-benzyl-4-piperidine, n-benzyl-4-piperidinone, n-benzyl-4-piperidone, n-benzylpiperidin-4-one |
PubChem CID | 19220 |
IUPAC Name | 1-benzylpiperidin-4-one |
SMILES | C1CN(CCC1=O)CC2=CC=CC=C2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC106131000
|
Thermo Fisher Scientific
106131000 |
100mL | Glass bottle |
Each for $139.21
|
|
AC106132500
|
Thermo Fisher Scientific
106132500 |
250mL | Glass bottle |
Each for $281.65
|
|
Chemical Identifiers
3612-20-2 | |
189.26 | |
SJZKULRDWHPHGG-UHFFFAOYSA-N | |
19220 | |
C1CN(CCC1=O)CC2=CC=CC=C2 |
C12H15NO | |
MFCD00006192 | |
1-benzyl-4-oxopiperidine, 1-benzyl-4-piperidinone, 1-benzyl-4-piperidone, 1-benzyl-piperidin-4-one, 1-benzylpiperidone, 4-piperidinone, 1-phenylmethyl, n-benzyl-4-piperidine, n-benzyl-4-piperidinone, n-benzyl-4-piperidone, n-benzylpiperidin-4-one | |
1-benzylpiperidin-4-one |
Specifications
1.0600g/mL | |
Authentic | |
1.5390 to 1.5410 | |
0.5% max. | |
Colorless to Straw | |
3612-20-2 | |
100.0 | |
C12H15NO | |
21,IV,3189 | |
Solubility in water: 12g/L water (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, d.m.f.,, acetone and chloroform | |
C1CN(CCC1=O)CC2=CC=CC=C2 | |
189.26 | |
189.26 | |
99% |
71°C | |
Glass bottle | |
1.06 | |
133.0°C to 135.0°C (7.0mmHg) | |
100mL | |
98.5 | |
98.5% min. (GC) | |
MFCD00006192 | |
1-benzyl-4-oxopiperidine, 1-benzyl-4-piperidinone, 1-benzyl-4-piperidone, 1-benzyl-piperidin-4-one, 1-benzylpiperidone, 4-piperidinone, 1-phenylmethyl, n-benzyl-4-piperidine, n-benzyl-4-piperidinone, n-benzyl-4-piperidone, n-benzylpiperidin-4-one | |
SJZKULRDWHPHGG-UHFFFAOYSA-N | |
1-benzylpiperidin-4-one | |
19220 | |
Oily Liquid | |
N-Benzyl-4-piperidone, 99% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.
GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 222-782-4
Recommended Storage : This product gains color during storage