missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Benzyl-4-piperidone, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 106131000

 View more versions of this product

Catalog No. AC106131000


 

Specifications Description & Specifications

Specifications

Oily Liquid
189.26
98 to 102%
3612-20-2
Authentic
98.5
N-Benzyl-4-piperidone
99%
1.5390 to 1.5410 (20°C, 589nm)
This product gains color during storage
Clear colorless to straw
100mL
100.0
1-(Phenylmethyl)-4-piperidinone
C12H15NO
MFCD00006192
99%
0.5% max.
Glass bottle

Chemical Identifiers

3612-20-2
100.0
C12H15NO
MFCD00006192
N-Benzyl-4-piperidone, 99%
98.5
99%
189.26
1-(Phenylmethyl)-4-piperidinone
98 to 102%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 222-782-4