missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyl-4-methylbenzamide, 97%

Manufacturer:  Alfa Aesar™ H5627003

 View more versions of this product

Catalog No. AAH5627003



Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. 

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Specifications

N-Benzyl-4-methylbenzamide
5436-83-9
MFCD00026008
QIULRTIURUDCGH-UHFFFAOYSA-N
N-benzyl-4-methylbenzamide
228681
97%
1g
C15H15NO
benzamide, 4-methyl-n-phenylmethyl, cambridge id 5104261, benzamide,4-methyl-n-phenylmethyl
CC1=CC=C(C=C1)C(=O)NCC2=CC=CC=C2
225.291
225.29

Chemical Identifiers

5436-83-9
225.291
QIULRTIURUDCGH-UHFFFAOYSA-N
228681
CC1=CC=C(C=C1)C(=O)NCC2=CC=CC=C2
C15H15NO
MFCD00026008
benzamide, 4-methyl-n-phenylmethyl, cambridge id 5104261, benzamide,4-methyl-n-phenylmethyl
N-benzyl-4-methylbenzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents