Alfa Aesar™ N-Benzyl-4-methoxybenzylamine, 97%
Manufacturer:  Alfa Aesar™ H55633MD
 View more versions of this product
Catalog No. AAH55633MD
Specifications
Specifications
N-Benzyl-4-methoxybenzylamine
14429-02-8
MFCD01002920
4-methoxybenzylbenzylamine
COC1=CC=C(C=C1)CNCC2=CC=CC=C2
227.307
227.31
250mg
C15H17NO
UN3259
WZUBICZIDUTTNJ-UHFFFAOYSA-N
N-[(4-methoxyphenyl)methyl]-1-phenylmethanamine
738564
97%
Chemical Identifiers
14429-02-8
227.307
WZUBICZIDUTTNJ-UHFFFAOYSA-N
738564
COC1=CC=C(C=C1)CNCC2=CC=CC=C2
C15H17NO
MFCD01002920
4-methoxybenzylbenzylamine
N-[(4-methoxyphenyl)methyl]-1-phenylmethanamine
Safety and Handling
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Documents