Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyl-4-methoxyaniline, 99%

Chemical Identifiers

Chemical Name or Material N-Benzyl-4-methoxyaniline
CAS 17377-95-6
Molecular Formula C14H15NO
Formula Weight 213.28
MDL Number MFCD00059298
UN Number UN2811
Beilstein 1877781
Synonym N-Benzyl-4-anisidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1115803

 
alfa aesar™
L1115803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1115806

 
alfa aesar™
L1115806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 213.28
Chemical Name or Material N-Benzyl-4-methoxyaniline
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Molecular Formula C14H15NO
MDL Number MFCD00059298
CAS 17377-95-6
UN Number UN2811
Boiling Point 138° to 140°C (0.1mmHg)
Quantity 1g
Synonym N-Benzyl-4-anisidine
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Assay 99%
Melting Point 48° to 50°C
Beilstein 1877781
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.