missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyl-4-fluorobenzylamine, 97%

Manufacturer:  Alfa Aesar™ H56362MD

 View more versions of this product

Catalog No. AAH56362MD



Description

Description

Applications
It is used as pharmaceutical intermediate.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Specifications

N-Benzyl-4-fluorobenzylamine
55096-88-3
MFCD00716882
SXZSRGKJZKOZRP-UHFFFAOYSA-N
N-[(4-fluorophenyl)methyl]-1-phenylmethanamine
784761
97%
250mg
C14H14FN
n-benzyl-1-4-fluorophenyl methanamine, benzyl-4-fluoro-benzyl-amine, benzyl-4-fluorobenzyl amine, benzyl 4-fluorophenyl methyl amine, benzyl 4-fluorobenzyl amine, 4-fluorophenyl methyl benzylamine, benzyl 4-fluorobenzyl amine, n-benzyl-n-4-fluorobenzyl amine, benzenemethanamine,4-fluoro-n-phenylmethyl, n-4-fluorophenyl methyl-1-phenylmethanamine
C1=CC=C(C=C1)CNCC2=CC=C(C=C2)F
215.271
215.27

Chemical Identifiers

55096-88-3
215.271
SXZSRGKJZKOZRP-UHFFFAOYSA-N
784761
C1=CC=C(C=C1)CNCC2=CC=C(C=C2)F
C14H14FN
MFCD00716882
n-benzyl-1-4-fluorophenyl methanamine, benzyl-4-fluoro-benzyl-amine, benzyl-4-fluorobenzyl amine, benzyl 4-fluorophenyl methyl amine, benzyl 4-fluorobenzyl amine, 4-fluorophenyl methyl benzylamine, benzyl 4-fluorobenzyl amine, n-benzyl-n-4-fluorobenzyl amine, benzenemethanamine,4-fluoro-n-phenylmethyl, n-4-fluorophenyl methyl-1-phenylmethanamine
N-[(4-fluorophenyl)methyl]-1-phenylmethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents