Please login to your online account to display your discounted pricing

N-Benzyl-4-cyano-4-(1-piperidino)-piperidine, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  325240050

 View more versions of this product

Catalog No. AC325240050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 283.42
Physical Form Crystalline Powder
Quantity 5g
Color Pale yellow
Melting Point 103° to 106°C
CAS Max % 100.0
Synonym 1'-(Phenylmethyl)-(1, 4'-bipiperidine)-4'-carbonitrile
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material N-Benzyl-4-cyano-4-(1-piperidino)-piperidine
CAS 84254-97-7
Assay Percent Range 98%
Molecular Formula C18H25N3
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
MDL Number MFCD00800635
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.

Specifications

Chemical Name or Material N-Benzyl-4-cyano-4-(1-piperidino)-piperidine
CAS 84254-97-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C18H25N3
Formula Weight 283.42
MDL Number MFCD00800635
Synonym 1'-(Phenylmethyl)-(1, 4'-bipiperidine)-4'-carbonitrile
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.

SDS & Safety Documents