missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyl-4-(chloromethyl)-N-phenylbenzamide, 97%

Chemical Identifiers

CAS 1858255-71-6
Molecular Formula C21H18ClNO
Molecular Weight (g/mol) 335.831
MDL Number MFCD22683138
InChI Key ITWJOMOXAADOGB-UHFFFAOYSA-N
Synonym n-benzyl-4-chloromethyl-n-phenylbenzamide
PubChem CID 98000580
IUPAC Name N-benzyl-4-(chloromethyl)-N-phenylbenzamide
SMILES C1=CC=C(C=C1)CN(C2=CC=CC=C2)C(=O)C3=CC=C(C=C3)CCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60582MD View Documents Alfa Aesar™
H60582MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6058203 View Documents Alfa Aesar™
H6058203
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1858255-71-6
335.831
ITWJOMOXAADOGB-UHFFFAOYSA-N
98000580
C1=CC=C(C=C1)CN(C2=CC=CC=C2)C(=O)C3=CC=C(C=C3)CCl
C21H18ClNO
MFCD22683138
n-benzyl-4-chloromethyl-n-phenylbenzamide
N-benzyl-4-(chloromethyl)-N-phenylbenzamide

Specifications

250mg
C21H18ClNO
n-benzyl-4-chloromethyl-n-phenylbenzamide
C1=CC=C(C=C1)CN(C2=CC=CC=C2)C(=O)C3=CC=C(C=C3)CCl
335.831
335.83
N-Benzyl-4-(chloromethyl)-N-phenylbenzamide
1858255-71-6
MFCD22683138
ITWJOMOXAADOGB-UHFFFAOYSA-N
N-benzyl-4-(chloromethyl)-N-phenylbenzamide
98000580
97%
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures