missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyl-4-(chloromethyl)-N-isopropylbenzamide, 97%

Chemical Identifiers

CAS 1097079-85-0
Molecular Formula C18H20ClNO
Molecular Weight (g/mol) 301.814
MDL Number MFCD13658458
InChI Key BLONTFYZTPFISN-UHFFFAOYSA-N
Synonym n-benzyl-4-chloromethyl-n-isopropylbenzamide
PubChem CID 43333466
IUPAC Name N-benzyl-4-(chloromethyl)-N-propan-2-ylbenzamide
SMILES CC(C)N(CC1=CC=CC=C1)C(=O)C2=CC=C(C=C2)CCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60184MD View Documents Alfa Aesar™
H60184MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6018403 View Documents Alfa Aesar™
H6018403
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1097079-85-0
301.814
BLONTFYZTPFISN-UHFFFAOYSA-N
43333466
CC(C)N(CC1=CC=CC=C1)C(=O)C2=CC=C(C=C2)CCl
C18H20ClNO
MFCD13658458
n-benzyl-4-chloromethyl-n-isopropylbenzamide
N-benzyl-4-(chloromethyl)-N-propan-2-ylbenzamide

Specifications

250mg
C18H20ClNO
n-benzyl-4-chloromethyl-n-isopropylbenzamide
CC(C)N(CC1=CC=CC=C1)C(=O)C2=CC=C(C=C2)CCl
301.814
301.81
N-Benzyl-4-(chloromethyl)-N-isopropylbenzamide
1097079-85-0
MFCD13658458
BLONTFYZTPFISN-UHFFFAOYSA-N
N-benzyl-4-(chloromethyl)-N-propan-2-ylbenzamide
43333466
97%