missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Benzyl-4-chlorobenzamide, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H55517MD

 View more versions of this product

Catalog No. AAH55517MD


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

N-Benzyl-4-chlorobenzamide
C14H12ClNO
250 mg
Soluble in water.
ClC1=CC=C(C=C1)C(=O)NCC1=CC=CC=C1
245.71
245.71
7461-34-9
MFCD00018682
n-benzyl-4-chloro-benzamide, 4-chloro-n-benzylbenzamide, n∼1∼-benzyl-4-chlorobenzamide, 4-chloro-n-phenylmethyl benzamide, 4-chlorophenyl-n-benzylcarboxamide, 4-chloranyl-n-phenylmethyl benzamide
LSMWDKIFKGLNSW-UHFFFAOYSA-N
N-benzyl-4-chlorobenzamide
346668
97%

Chemical Identifiers

7461-34-9
245.71
LSMWDKIFKGLNSW-UHFFFAOYSA-N
346668
ClC1=CC=C(C=C1)C(=O)NCC1=CC=CC=C1
C14H12ClNO
MFCD00018682
n-benzyl-4-chloro-benzamide, 4-chloro-n-benzylbenzamide, n∼1∼-benzyl-4-chlorobenzamide, 4-chloro-n-phenylmethyl benzamide, 4-chlorophenyl-n-benzylcarboxamide, 4-chloranyl-n-phenylmethyl benzamide
N-benzyl-4-chlorobenzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions