missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyl-4-chlorobenzamide, 97%

Manufacturer:  Alfa Aesar™ H5551703

 View more versions of this product

Catalog No. AAH5551703



Description

Description

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

N-Benzyl-4-chlorobenzamide
7461-34-9
MFCD00018682
Soluble in water.
C1=CC=C(C=C1)CNC(=O)C2=CC=C(C=C2)Cl
245.706
245.71
1g
C14H12ClNO
4-chloranyl-n-phenylmethyl benzamide
LSMWDKIFKGLNSW-UHFFFAOYSA-N
N-benzyl-4-chlorobenzamide
346668
97%

Chemical Identifiers

7461-34-9
245.706
LSMWDKIFKGLNSW-UHFFFAOYSA-N
346668
C1=CC=C(C=C1)CNC(=O)C2=CC=C(C=C2)Cl
C14H12ClNO
MFCD00018682
4-chloranyl-n-phenylmethyl benzamide
N-benzyl-4-chlorobenzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents