missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyl-4-chloro-N-phenylbenzamide, 97%

$211.27 - $675.45

Chemical Identifiers

CAS 701918-26-5
Molecular Formula C20H16ClNO
Molecular Weight (g/mol) 321.804
MDL Number MFCD06251246
InChI Key LDDXRNJXGXWUQJ-UHFFFAOYSA-N
Synonym 4-chlorophenyl-n-phenyl-n-benzylcarboxamide
PubChem CID 20787883
IUPAC Name N-benzyl-4-chloro-N-phenylbenzamide
SMILES C1=CC=C(C=C1)CN(C2=CC=CC=C2)C(=O)C3=CC=C(C=C3)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59845MD View Documents Alfa Aesar™
H59845MD
250mg Each for $211.27
 
AAH5984503 View Documents Alfa Aesar™
H5984503
1g Each for $675.45
 
Specifications

Chemical Identifiers

701918-26-5
321.804
LDDXRNJXGXWUQJ-UHFFFAOYSA-N
20787883
C1=CC=C(C=C1)CN(C2=CC=CC=C2)C(=O)C3=CC=C(C=C3)Cl
C20H16ClNO
MFCD06251246
4-chlorophenyl-n-phenyl-n-benzylcarboxamide
N-benzyl-4-chloro-N-phenylbenzamide

Specifications

250mg
C20H16ClNO
4-chlorophenyl-n-phenyl-n-benzylcarboxamide
C1=CC=C(C=C1)CN(C2=CC=CC=C2)C(=O)C3=CC=C(C=C3)Cl
321.804
321.81
N-Benzyl-4-chloro-N-phenylbenzamide
701918-26-5
MFCD06251246
LDDXRNJXGXWUQJ-UHFFFAOYSA-N
N-benzyl-4-chloro-N-phenylbenzamide
20787883
97%