missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Benzyl-2-naphthylamine, TCI America™

$311.67 - $1,077.11

Chemical Identifiers

CAS 13672-18-9
Molecular Formula C17H15N
Molecular Weight (g/mol) 233.314
MDL Number MFCD00144848
InChI Key KBOITYDYBXNTMF-UHFFFAOYSA-N
PubChem CID 12487301
IUPAC Name N-benzylnaphthalen-2-amine
SMILES C1=CC=C(C=C1)CNC2=CC3=CC=CC=C3C=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B30885G View Documents TCI America
B30885G
5g Each for $311.67
 
B308825G View Documents TCI America
B308825G
25g Each for $1,077.11
 
Specifications

Chemical Identifiers

13672-18-9
233.314
KBOITYDYBXNTMF-UHFFFAOYSA-N
N-benzylnaphthalen-2-amine
C17H15N
MFCD00144848
12487301
C1=CC=C(C=C1)CNC2=CC3=CC=CC=C3C=C2

Specifications

190°C
68°C
13672-18-9
MFCD00144848
C1=CC=C(C=C1)CNC2=CC3=CC=CC=C3C=C2
233.314
233.31
≥98.0% (GC)
Undesignated
5g
C17H15N
KBOITYDYBXNTMF-UHFFFAOYSA-N
N-benzylnaphthalen-2-amine
12487301
Crystal-Powder at 20°C
N-Benzyl-2-naphthylamine
Safety and Handling

Safety and Handling

TSCA : No