Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyl-2-chloronicotinamide, 96%

$267.34 - $1,323.35

Chemical Identifiers

Chemical Name or Material N-Benzyl-2-chloronicotinamide
CAS 65423-28-1
Molecular Formula C13H11ClN2O
Formula Weight 246.70g/mol
MDL Number MFCD00831758
Synonym N-Benzylpyridine-3-carboxamide; 2-Chloronicotinic acid benzylamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5015203

 
alfa aesar™
H50152-03
Each for $267.34

AAH5015206

 
alfa aesar™
H50152-06
Each for $1,323.35
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material N-Benzyl-2-chloronicotinamide
CAS 65423-28-1
Assay 96%
Molecular Formula C13H11ClN2O
Formula Weight 246.70g/mol
MDL Number MFCD00831758
Synonym N-Benzylpyridine-3-carboxamide; 2-Chloronicotinic acid benzylamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number QS4030000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.