missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyl-2-chloro-N-phenylbenzamide, 97%

$211.27 - $675.45

Chemical Identifiers

CAS 500873-57-4
Molecular Formula C20H16ClNO
Molecular Weight (g/mol) 321.804
MDL Number MFCD03731396
InChI Key OWBLYVPLNRYXED-UHFFFAOYSA-N
PubChem CID 295307
IUPAC Name N-benzyl-2-chloro-N-phenylbenzamide
SMILES C1=CC=C(C=C1)CN(C2=CC=CC=C2)C(=O)C3=CC=CC=C3Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59901MD View Documents Alfa Aesar™
H59901MD
250mg Each for $211.27
 
AAH5990103 View Documents Alfa Aesar™
H5990103
1g Each for $675.45
 
Specifications

Chemical Identifiers

500873-57-4
321.804
OWBLYVPLNRYXED-UHFFFAOYSA-N
N-benzyl-2-chloro-N-phenylbenzamide
C20H16ClNO
MFCD03731396
295307
C1=CC=C(C=C1)CN(C2=CC=CC=C2)C(=O)C3=CC=CC=C3Cl

Specifications

250mg
C20H16ClNO
OWBLYVPLNRYXED-UHFFFAOYSA-N
N-benzyl-2-chloro-N-phenylbenzamide
295307
97%
500873-57-4
MFCD03731396
C1=CC=C(C=C1)CN(C2=CC=CC=C2)C(=O)C3=CC=CC=C3Cl
321.804
321.81
N-Benzyl-2-chloro-N-phenylbenzamide