missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyl-2,6-difluoro-N-phenylbenzamide, 97%

Chemical Identifiers

CAS 1426315-84-5
Molecular Formula C20H15F2NO
Molecular Weight (g/mol) 323.343
MDL Number MFCD22683100
InChI Key GPQFSRTYCAFMCY-UHFFFAOYSA-N
PubChem CID 98000094
IUPAC Name N-benzyl-2,6-difluoro-N-phenylbenzamide
SMILES C1=CC=C(C=C1)CN(C2=CC=CC=C2)C(=O)C3=C(C=CC=C3F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59925MD View Documents Alfa Aesar™
H59925MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5992503 View Documents Alfa Aesar™
H5992503
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1426315-84-5
323.343
GPQFSRTYCAFMCY-UHFFFAOYSA-N
N-benzyl-2,6-difluoro-N-phenylbenzamide
C20H15F2NO
MFCD22683100
98000094
C1=CC=C(C=C1)CN(C2=CC=CC=C2)C(=O)C3=C(C=CC=C3F)F

Specifications

250mg
C20H15F2NO
GPQFSRTYCAFMCY-UHFFFAOYSA-N
N-benzyl-2,6-difluoro-N-phenylbenzamide
98000094
97%
1426315-84-5
MFCD22683100
C1=CC=C(C=C1)CN(C2=CC=CC=C2)C(=O)C3=C(C=CC=C3F)F
323.343
323.34
N-Benzyl-2,6-difluoro-N-phenylbenzamide