Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(Benzoylmethyl)pyridinium bromide, 98%

$31.09 - $105.58

Chemical Identifiers

Chemical Name or Material N-(Benzoylmethyl)pyridinium bromide
CAS 16883-69-5
Molecular Formula C13H12BrNO
Formula Weight 278.15
MDL Number MFCD00031995
Beilstein 3599124
Synonym N-Phenacylpyridinium bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2243709

 
alfa aesar™
B2243709
10g Each for $31.09

AAB2243718

 
alfa aesar™
B2243718
50g Each for $105.58
Description & Specifications

Specifications

Formula Weight 278.15
Chemical Name or Material N-(Benzoylmethyl)pyridinium bromide
Assay 98%
Molecular Formula C13H12BrNO
Melting Point 193° to 195°C
MDL Number MFCD00031995
CAS 16883-69-5
Beilstein 3599124
Quantity 10g
Synonym N-Phenacylpyridinium bromide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.