missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzoyl-N-phenylhydroxylamine, 98%

Manufacturer:  Alfa Aesar™ A1195014

 View more versions of this product

Catalog No. AAA1195014



Specifications

Specifications

N-Benzoyl-N-phenylhydroxylamine
25g
C13H11NO2
2212449
YLYIXDZITBMCIW-UHFFFAOYSA-N
N-hydroxy-N-phenylbenzamide
67536
98%
118°C to 120°C
304-88-1
MFCD00002111
n-benzoyl-n-phenylhydroxylamine, n-phenylbenzohydroxamic acid, n-hydroxybenzanilide, benzamide, n-hydroxy-n-phenyl, n-benzoyl-n-phenylhydroxamic acid, n-benzoylphenylhydroxylamine, benzohydroxamic acid, n-phenyl, tantalon, acmc-1cqm9, n-phenylbenzohydroxamicacid
C1=CC=C(C=C1)C(=O)N(C2=CC=CC=C2)O
213.236
213.24

Chemical Identifiers

304-88-1
213.236
YLYIXDZITBMCIW-UHFFFAOYSA-N
67536
C1=CC=C(C=C1)C(=O)N(C2=CC=CC=C2)O
C13H11NO2
MFCD00002111
n-benzoyl-n-phenylhydroxylamine, n-phenylbenzohydroxamic acid, n-hydroxybenzanilide, benzamide, n-hydroxy-n-phenyl, n-benzoyl-n-phenylhydroxamic acid, n-benzoylphenylhydroxylamine, benzohydroxamic acid, n-phenyl, tantalon, acmc-1cqm9, n-phenylbenzohydroxamicacid
N-hydroxy-N-phenylbenzamide
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents