Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzoyl-L-phenylalanine, 98%

Chemical Identifiers

Chemical Name or Material N-Benzoyl-L-phenylalanine
CAS 2566-22-5
Formula Weight 269.3
MDL Number MFCD00063153
Synonym Bz-Phe-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6660806

 
alfa aesar™
H6660806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6660814

 
alfa aesar™
H6660814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 269.3
Chemical Name or Material N-Benzoyl-L-phenylalanine
CAS 2566-22-5
Physical Form Powder
Assay 98%
Synonym Bz-Phe-OH
Color Off-white to white
Quantity 5g
RTECS Number SQ7326700
Storage Note 1 2° to 8°C
MDL Number MFCD00063153
Safety and Handling

2° to 8°C