Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzoyl-L-glutamic acid, 98%

Chemical Identifiers

Chemical Name or Material N-Benzoyl-L-glutamic acid
CAS 6094-36-6
Formula Weight 251.24
MDL Number MFCD00014028
Beilstein 2622989
Synonym Bz-Glu-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6677806

  SDS
alfa aesar™
H6677806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6677814

  SDS
alfa aesar™
H6677814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6677822

  SDS
alfa aesar™
H6677822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 251.24
Physical Form Solid
Assay 98%
MDL Number MFCD00014028
CAS 6094-36-6
Beilstein 2622989
Chemical Name or Material N-Benzoyl-L-glutamic acid
Synonym Bz-Glu-OH
Quantity 5g