Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzoyl-L-alanine, 95%

Chemical Identifiers

Chemical Name or Material N-Benzoyl-L-alanine
CAS 2198-64-3
Formula Weight 193.2
MDL Number MFCD00063128
Synonym Bz-Ala-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6669706

 
alfa aesar™
H6669706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6669714

 
alfa aesar™
H6669714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6669722

 
alfa aesar™
H6669722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 193.2
Chemical Name or Material N-Benzoyl-L-alanine
Physical Form Solid
Assay 95%
MDL Number MFCD00063128
CAS 2198-64-3
Synonym Bz-Ala-OH
Storage Note 1 -30° to -10°C
Quantity 5g
Safety and Handling

-30° to -10°C