missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Benzoyl-DL-alanine, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1310009
Description
N-Benzoyl-DL-alanine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Benzoyl-DL-alanine is used as pharmaceutical intermediate.
Solubility
Very slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
163°C to 167°C | |
1205-02-3 | |
MFCD00020393 | |
n-benzoyl-dl-alanine, benzoyl-dl-alanine, methylhippuric acid, n-benzoylalanine, alanine, n-benzoyl, benzoylalanine, dl-n-benzoylalanine, 2-phenylformamido propanoic acid, alpha-methylhippuric acid, n-phenylcarbonyl alanine | |
UAQVHNZEONHPQG-UHFFFAOYNA-N | |
2-benzamidopropanoic acid | |
71002 | |
193.2 | |
10 g |
N-Benzoyl-DL-alanine | |
C10H11NO3 | |
3201778 | |
Very slightly soluble in water. | |
CC(NC(=O)C1=CC=CC=C1)C(O)=O | |
193.20 | |
CHEBI:71167 | |
≥97% |
Chemical Identifiers
1205-02-3 | |
193.20 | |
UAQVHNZEONHPQG-UHFFFAOYNA-N | |
71002 | |
2-benzamidopropanoic acid |
C10H11NO3 | |
MFCD00020393 | |
n-benzoyl-dl-alanine, benzoyl-dl-alanine, methylhippuric acid, n-benzoylalanine, alanine, n-benzoyl, benzoylalanine, dl-n-benzoylalanine, 2-phenylformamido propanoic acid, alpha-methylhippuric acid, n-phenylcarbonyl alanine | |
CHEBI:71167 | |
CC(NC(=O)C1=CC=CC=C1)C(O)=O |
Safety and Handling
EINECSNumber : 214-879-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only