Please login to your online account to display your discounted pricing

N-Arachidonylglycine, Tocris

Novel endocannabinoid; Suppresses pain in vivo

$146.74 - $618.87

Chemical Identifiers

Chemical Name or Material N-Arachidonylglycine
Assay Percent Range >98%
Molecular Formula C22H35NO3
Formula Weight 361.52
Synonym N-(1-oxo-5Z,8Z,11Z,14Z-eicosatetraenyl)glycine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

144525

 
r & d systems
RDS144525
25mg Each for $618.87

14455

  SDS
r & d systems
RDS14455
5mg Each for $146.74
Description & Specifications

Specifications

Chemical Name or Material N-Arachidonylglycine
CAS 179113-91-8
Recommended Storage Desiccate at -20°C
Assay Percent Range >98%
Molecular Formula C22H35NO3
Formula Weight 361.52
Synonym N-(1-oxo-5Z,8Z,11Z,14Z-eicosatetraenyl)glycine
Quantity 25mg
Safety and Handling

Desiccate at -20°C