Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Arachidonylglycine

Chemical Identifiers

Chemical Name or Material N-Arachidonylglycine
CAS 179113-91-8
Formula Weight 361.52
MDL Number MFCD03791313
Synonym AA-Gly
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67492LB0

 
alfa aesar™
J67492LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 361.52
Chemical Name or Material N-Arachidonylglycine
CAS 179113-91-8
Synonym AA-Gly
MDL Number MFCD03791313
Physical Form Waxy Solid
Quantity 5mg
Color White
Sensitivity Air Sensitive
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C