Please login to your online account to display your discounted pricing

N-ArachidonylGABA, Tocris

Inhibits pain in vivo

$146.74 - $618.87

Chemical Identifiers

Chemical Name or Material N-ArachidonylGABA
Assay Percent Range >98%
Molecular Formula C24H39NO3
Formula Weight 389.58
Synonym 4-[[(5Z,8Z,11Z,14Z)-1-Oxo-5,8,11,14-eicosatetraenyl]amino]butanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

181425

 
r & d systems
RDS181425
25mg Each for $618.87

18145

 
r & d systems
RDS18145
5mg Each for $146.74
Description & Specifications

Specifications

Formula Weight 389.58
Quantity 25mg
Assay Percent Range >98%
Synonym 4-[[(5Z,8Z,11Z,14Z)-1-Oxo-5,8,11,14-eicosatetraenyl]amino]butanoic acid
CAS 128201-89-8
Chemical Name or Material N-ArachidonylGABA
Molecular Formula C24H39NO3
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C