Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Arachidonylcyclopropylamide, 98%


Quantity 10mg

Manufacturer: alfa aesar™  J67376MA

 View more versions of this product

Catalog No. AAJ67376MA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 343.55
Chemical Name or Material N-Arachidonylcyclopropylamide
Assay 98%
CAS 229021-64-1
Sensitivity Air Sensitive
Physical Form Liquid
MDL Number MFCD02683582
Synonym ACPA
Color Pale yellow
Quantity 10mg
Storage Note 1 -30° to -10°C
RTECS Number JX3841600

Specifications

Chemical Name or Material N-Arachidonylcyclopropylamide
CAS 229021-64-1
Formula Weight 343.55
MDL Number MFCD02683582
Synonym ACPA
Accessories

-30° to -10°C